Learn More
Chemsplash Eka Trouser
GREENER_CHOICE
Codice prodotto. 30058358
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Dimensione della confezione:
pezzi 50
Codice prodotto. 30058358
Marca: Chemsplash 2616.400.005
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Trousers with Elasticated Waist and Ankles are strong and resistant to chemical splash and dust.
Chemsplash Trousers with Elasticated Waist and Ankles
  • Chemsplash 55GSM Type PB 6B Fabric
  • Elasticated Waist and Ankles
  • Silicone and Latex Free
  • Anti-Static Material
  • Non-Linting Fabric
  • PFAS Free
  • Conforms to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9
Description: Chemsplash Trousers with Elasticated Waist and Ankles are made of soft and microporous Chemsplash fabric. They are strong and resistant to chemical splash and dust, lint-free, and feature anti-static properties under EN1149-5:2008. Chemsplash accessories are designed to protect the wearer from the environment or the environment from the wearer. This range has been crafted to offer maximum comfort and wearability. The trousers conform to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9 standards.
Specifica
Size Medium
Sesso Unisex
Product Type Trouser
Materiale Multi-layer Barrier Laminate Fabric
Colore White
Ankle Style Elastic
Tipo di giuntura Stitched
N. di tasche None
Monouso Single-use
Senza lattice Yes
Sterilità Non-sterile