Learn More
Thermo Scientific™ ELISA Systems™ Food Allergen Swab Kit for the use with ELISA Systems™ Food Allergen Test Kits.
Descrizione
Simplify food allergen testing in food and environmental samples with ELISA Systems™ Food Allergen Swab Kit. The kit comes with sterile swabs and specially formulated, sterile swab wetting solution for each swab.
When used appropriately, they can provide detailed information regarding the presence of detectable allergenic material. This is of particular importance in environmental monitoring situations, such as:
- Verification of sanitation procedures
- Raw material storage facilities
- Food preparation facilities
The ELISA Systems Swab Residue Kit is designed to provide a convenient supply of swab items for use in the collection of swab samples.
Not all products are available for sale in all territories. Please inquire.ELISA Systems™ Food Allergen Test Kits are manufactured by ELISA Systems Pty Ltd.
ELISA Systems is a trademark of ELISA Systems Pty Ltd.
Specifica
Specifica
| Descrizione | ELISA Systems Food Allergen Swab Kit |
| Immunoassay Kit Format | ELISA (Standard) |
| Product Type | Food Allergen Swab Kit |
| Contenuto kit | 10 x sterile swab wetting solution, 10 x sterile swabs |
| Quantità | 10 Pk. |
| Requisiti di stoccaggio | 20°C to 25°C |