Learn More
Thermo Scientific™ Elite pH Pocket Testers and Replacement Sensors
Codice prodotto. p-7139301
Click to view available options
Dimensione della confezione:
Pezzo
Product Type:
pH Tester
Codice prodotto. 16846966
Marca: Thermo Scientific™ 01X001535
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Flexible on-the-go pH and temperature testing using durable Elite pocket testers with replaceable sensors for a variety of applications including environmental monitoring, pools, spas and more!
Benefit from these pocket-sized testers. Thermo Scientific™ Elite testers are designed to provide on-the-go measurements for a variety of applications including homebrew beer and wine, food and beverage, hydroponics, aquaculture and aquariums, agriculture, pools and spas, environmental monitoring, education and more. The Elite CTS cup-sensor pocket testers are ideal for flexible on-the-go conductivity, TDS, salinity and temperature testing in a compact, travel-ready design.
Elite Pocket Testers offer flexible on-the-go electrochemical testing. Choose the Elite pH model to measure pH with temperature using a direct-measure sensor design for your application. Elite pocket testers are durable, with a large backlit screen, one-touch calibration feature and replaceable sensor heads. They are ideal for testing beverages including homebrew beer and wine, food, hydroponics, aquaculture and aquariums, environmental monitoring and more.
  • Flexible testing option
  • Measure pH and temperature
  • Large back-lit graphic display
  • Uses 4 x AAA alkaline batteries
  • Average 250 hours of battery life
  • Wide leak-proof cap serves as stable base
  • Replaceable sensors
  • One-year warranty for tester
  • Six-month warranty for replacement sensors
TRUSTED_SUSTAINABILITY
Specifica
Descrizione Replacement sensor for Elite pH pocket tester
Include 1 replacement pH sensor
Precisione (Temperatura) ±0.6°C/±1.0°F for 0.0deg;C to 50.0°C
Precisione (pH) ±0.02
Measurement Modes pH, Temperature
Intervallo (pH) -1 to 15
Risoluzione (pH) 0.01
Temperature Range (Metric) 0°C to 60°C
Intervallo di temperatura (sistema imperiale) 32.0°F to 140.0°F
Risoluzione (temperatura) 0.1°C, 0.1°F
Calibrazione della sonda di temperatura 1 point
Warranty 6 Months for Replacement Sensor
Intervallo umidità relativa 5 to 85 %, non-condensing
Compensazione della temperatura Yes
Temperatura (sistema metrico) operativa 5°C to 45°C (ambient)
Tipo Replacement pH Sensor