Learn More
Merck Fluid Contamination Monitors
Codice prodotto. 11596465 Sfoglia Tutto Merck Prodotti
Click to view available options
Tipo:
Fluid Contamination Monitor
Matched Weight Monitor
Dimensione della confezione:
pezzi 50
Codice prodotto. 11596465
Marca: Merck MAWP037PM
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Fluid Contamination Analysis Monitors are transparent, disposable filter holders made from Tenite™ plastic and pre-assembled with MF-Millipore filters (effective filtration area is 9 cm2).
Transparent, disposable filter holders made from Tenite™ plastic and pre-assembled with MF-Millipore filters
A thick cellulose pad, 34 mm diameter, between the monitor sections seals the filter to distribute sample flow evenly over the filter surface. Use with Fluid Sampling Kit (XX6403730).Matched-Weight Monitors:Matched-Weight Monitors eliminate test filter pre-weighing in gravimetric analysis. Each monitor contains two filters matched in weight to within 0.1 mg. Sample passes through both, but upper (test) filter retains all contaminant. The difference in filter weights, after drying, equals the weight of contaminant in sample.
TRUSTED_SUSTAINABILITY
Specifica
Materiale Tenite Plastic
Dimensione del poro 0.8 μm
Sterilità Non-sterile
Area di filtraggio 9 cm2
Da utilizzare con (applicazione) Gravimetric particle analysis
Tipo Matched Weight Monitor
Diameter (English) Inside 1.45 in.
Diameter (Metric) Inside 3.7 cm