Learn More
Merck Vacuum Tubing
Codice prodotto. 10527841 Sfoglia Tutto Merck Prodotti
Click to view available options
Materiale:
Latex
Silicone
Dimensione della confezione:
Pezzo
Codice prodotto. 10527841
Marca: Merck XX6700037
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Accessori per portafiltri per la preparazione dei campioni.
Millipore has a broad selection of fittings, adapters, valves, gauges, and tubing that can be used in conjunction with Millipore filtration systems. With stated exceptions, material of construction is Series 300 stainless steel.Tri-Clamp (TC) fittings are referred to by convention as 1 1/2 in., 2 in., etc. 1 1/2 in. TC fittings connect to 1 in. or 1 1/2 in. O.D. tube sizes. 2 in. TC fittings connect to 2 in. O.D. tube size only.
TRUSTED_SUSTAINABILITY
Specifica
Tipo Laboratory Filter Holders or Cyclones
Overall Length (English) 10 ft.
Lunghezza complessiva (sistema metrico) 305 cm
Materiale Latex