Learn More
Merck Millicell™ Culture Plate Inserts: HATF
Sfoglia Tutto Merck Prodotti
Click to view available options
Diametro filtro (sistema metrico):
12mm
30mm
Altezza (sistema metrico):
10.5mm
13mm
Dimensione della confezione:
pezzi 50
Descrizione
- Microporous membranes allow access to cell monolayer from both apical and basolateral domains; promote enhanced cell attachment, growth, and differentiation of attachment-dependent and suspension cultures
- Ready-to-use Milicell inserts fit most 6- or 24-well plates and are easily prepared for SEM and TEM techniques; compatible with standard and fluorescent stains
- Effective growth area for 12mm diameter inserts, 0.6cm2; and 4.2cm2 for 30mm diameter inserts
- Don't require an extracellular matrix for attachment-dependent cells
- Polystyrene housing; supplied sterile and individually blister packed
- Inserts utilize a mixed cellulose esters membrane; Triton™-free
Applications:
Transport and permeability studies, co-culture for cell-cell interaction studies, in vitro toxicity testing, tumor cell metastasis studies, and organotypic cultures
Specifica
Specifica
| Area di coltura | 0.6 cm2 |
| Dimensione del poro | 0.45 μm |
| Da utilizzare con (apparecchiatura) | 24 Well Plate |
| Diametro filtro (sistema metrico) | 12mm |
| Altezza (sistema metrico) | 10.5mm |
| Sterilità | Sterile |
Titolo del prodotto
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering