Descrizione
Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining
- Membrane-to-plate seal ensures no reagent crosstalk and limited optical crosstalk Patented underdrain support structure allows liquid volumes to be maintained for incubation times of up to 21 days
Specifica
Specifica
| Formato | Standard |
| Coperchio | Yes |
| Membrana | PVDF idrofobo |
| Matrice | 8 x 12 |
| Dimensione del poro | 0.45 μm |
| Product Line | MultiScreen |
| Colore | Trasparente |
| Product Type | MultiScreen-IP with Underdrain |
| Materiale | Acrilico |
| No. of Wells | 96 |
| Vedi altri risultati |