Learn More
Merck Samplicity™ Filtration System Vial Trays
Sfoglia Tutto Merck Prodotti
Click to view available options
Tipo:
Vial Trays
Waste Trays
Materiale:
Polypropylene
Recycled Polyester
Dimensione della confezione:
pezzi 2
pezzi 5
Descrizione
Overview of Samplicity™ G2 System
- Samplicity™ G2 Filtration System enables vacuum driven filtration of up to 8 samples at once, directly into HPLC vials.
- Filtration for HPLC sample prep without syringes. Sample is added to an adapter funnel on a standard Millex™ 33 mm filter (Millex™ Filters for the Samplicity™ G2 System) or an integrated funnel and filter (Millex Samplicity™ filter)
- Samplicity™ G2 System has lower ergonomic impact than manual syringe filtration
Applications: HPLC; Filtration of viscous or particulate laden samples for HPLC
Specifica
Specifica
| Da utilizzare con (apparecchiatura) | Samplicity G2 Filtration System |
| Materiale | Polypropylene |
| Sterilità | Non-sterile |
| Da utilizzare con (applicazione) | Analytical sample prep, High Performance Liquid Chromatography (HPLC) |
| N. per confezione | 2 Trays |
| Tipo | Vial Trays |