Learn More
Ansell ENCORE™ Polyisoprene and Neoprene Hybrid Surgical Gloves
Codice prodotto. 17833692
Click to view available options
Size:
5.5
6
6.5
7
7.5
8
8.5
9
Dimensione della confezione:
50 paia
Codice prodotto. 17833692
Marca: Ansell 340105055
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Polyisoprene and neoprene surgical gloves blend comfort and durability when using as an outer glove during double gloving.
  • ASTM D6978, ASTM F739 certified
  • HYBRID Technology blend of polyisoprene and neoprene offers the best of both materials, combining durability with latex-like comfort
  • Natural, semi-transparent glove, ideal to be used as an outer glove when double gloving with a colored underglove for fast and easy glove breach detection
  • Non-latex to eliminate the risk of latex Type I allergy
  • Free of zinc 2-mercaptobenzothiazole, diphenylguanidine, and cetylpyridinium chloride to minimize risk of chemical Type IV allergies and sensitivities to these chemical accelerators
Specifica
Certificazione/Conformità ASTM D6978, ASTM F739, ISO 10282
Certifications/Compliance (Europe) EN 16523-1, EN 374-2 and -4, EN 420:2003 + A1:2009, EN 455 1-4, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-1:2016, EN ISO 374-5:2016, EN ISO 374-5:2016
Accelerator Free No
Applicazioni Healthcare
AQL 0.65
Colore Natural
Monouso Single-use
Finitura esterna Partially Textured
Finger Thickness (English) 8.27 ±1.18 mil
Finger Thickness (Metric) 0.21 ±0.03 mm
Hand Specific
Laboratory/Cleanroom Use No
Latex Free
Materiale Polyisoprene, Neoprene
Palm Thickness (English) 7.48 ±1.18 mil
Palm Thickness (Metric) 0.19 ±0.03 mm
Powdered Powder Free
Size 5.5
Sterilità Sterile
Spessore (sistema imperiale) 7.48 ±1.18 mil
Spessore (sistema metrico) 0.19 ±0.03 mm
Modello ENCORE-340105
Lunghezza (sistema imperiale) 12 in.
Lunghezza (sistema metrico) 306 mm