Learn More
ENMD-1198, MedChemExpress
Product Code. 16481599
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481599 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16481599 Supplier Medchem Express Supplier No. HY16196
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress ENMD-1198 (IRC-110160), an orally active microtubule destabilizing agent, is a 2-methoxyestradiol analogue with antiproliferative and antiangiogenic activity. ENMD-1198 is suitable for inhibiting HIF-1alpha and STAT3 in human HCC cells and leads to reduced tumor growth and vascularization.
ENMD-1198 (IRC-110160), an orally active microtubule destabilizing agent, is a 2-methoxyestradiol analogue with antiproliferative and antiangiogenic activity. ENMD-1198 is suitable for inhibiting HIF-1alpha and STAT3 in human HCC cells and leads to reduced tumor growth and vascularization.
Specifica
Chemical Name or Material ENMD-1198
CAS 864668-87-1
Color Brownish Yellow
Purity Grade Notes Research
Molecular Formula C20H25NO2
Quantity 1 mg
Synonym IRC-110160
SMILES C[C@@]12C=CC[C@@]1([H])[C@]3([H])CCC4=C(C=C(OC)C(C(N)=O)=C4)[C@@]3([H])CC2
Molecular Weight (g/mol) 311.42
Formula Weight 311.42
Percent Purity 98.87%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Research purposes only