Learn More
Sartorius Entris™ II Precision Balance, Internal Adjustment
Codice prodotto. p-7228466 Sfoglia Tutto Sartorius Prodotti
Click to view available options
Capacità:
220 g
320 g
420 g
620 g
650 g
820 g
1200 g
2200 g
3200 g
4200 g
5200 g
6200 g
8200 g
Leggibilità:
1 mg
10 mg
100 mg
1000 mg
Modello:
CEU Electronic Toploading Balance
Dimensione della confezione:
1 unità
Codice prodotto. 16354698
Marca: Sartorius BCE223I1CEU
Entris™ II Precision Balance with Internal Adjustment offering isoCAL, LED touch technology, and 12 built-in applications at a budget price. Every balance offers quality, value and consistency.
  • No matter what you're weighing, the new Entris™ II balance is always the right choice.
  • Offering unrivalled value and backed by almost 150 years of German engineering expertise, the Entris™ II comes in two product lines, so you can find the balance that meets your specific weighing needs.
  • With over 40 models from which to choose, you will surely find a model to meet your basic weighing needs.
TRUSTED_SUSTAINABILITY
Specifica
Certificazione/Conformità CEU
Tipo di collegamento RS232, USB C
Capacità 220 g
Tipo di display Touchscreen
Leggibilità 1 mg
Materiale PBT, Glass, Stainless Steel
Modello CEU Electronic Toploading Balance
Tempo di stabilizzazione ≤1.5 sec.
Tipo Precision Balance
Requisiti elettrici 100 – 240 VAC (±10%), 50 - 60 Hz, 0.2 A