missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EOMES Mouse anti-Human, Mouse, PE, Clone: X4-83, BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 566749
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
EOMES Monoclonal antibody specifically detects EOMES in Human, Mouse samples. It is validated for Flow Cytometry
Specifica
| EOMES | |
| Monoclonal | |
| 0.2 mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 0.1 mg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed. |
| Flow Cytometry | |
| X4-83 | |
| PE | |
| C77258; T-box brain protein 2; TBR-2; TBR2; Tbr2; eomesodermin homolog | |
| Human EOMES aa 1-275 Recombinant Protein | |
| RUO | |
| Human, Mouse | |
| IgG1 κ |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto