Learn More
EPHA2 Mouse anti-Human, Unconjugated, Clone: 1A9C3, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66736-1-IG-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The EPHA2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EPHA2. This antibody recognizes human antigen. The EPHA2 antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifica
| EPHA2 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Flow Cytometry | |
| 1A9C3 | |
| Unconjugated | |
| 150 μL | |
| 1969 | |
| IgG1 |