Learn More
Fisherbrand™ Epoxy Corps Amber pH Electrode
Click to view available options
Tipo:
Electrode
Dimensione della confezione:
Pezzo
Descrizione
- Double junction reference is recommended for samples containing organic compounds, proteins, heavy metals or compounds that interact with silver (e.g. sulfide, bromide, iodide)
- All electrodes are available with a detachable lead or with a cable/connector attached
- Designed for all existing Brinkmann, Metrohm and Schott titration systems
- The electrode body is constructed with a glass ground joint that will fit their titration vessels
- Ionics and Tytronics titrators can be fitted with this same electrode without the ground joint
- Either design can be ordered depending on the titration unit
Specifica
Specifica
| Descrizione | Epoxy Corps Amber pH Electrode |
| Tipo | Electrode |