Learn More
Eppendorf™ HeatSealer S200 Accessory, Plate Adapters
Product Code. 15743155 Shop All Eppendorf Products
Click to view available options
Product Type:
High Profile Adapter
Low Profile Adapter
PCR Plate Adapter
Unit Size:
Pezzo
Product Code. 15743155
Brand: Eppendorf™ 5392070020
For use with HeatSealer S200
  • Hermetic sealing of multiwell plates
  • Eliminates evaporation in PCR, reducing cross contamination
  • Seals 96- and 384-well plates
  • Integrated thermostat prevents overheating
  • Improved mechanics for even easier sealing
  • PCR Plate Adapter included
TRUSTED_SUSTAINABILITY
Specifications
Wattage 350 W
Da utilizzare con (apparecchiatura) HeatSealer S200
Intervallo temperatura sigillatura 125°C to 200°C in 1°C increments
Product Type Low Profile Adapter