missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERp72 Mouse anti-Human, Clone: 1D5F3, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66365-1-IG-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
ERp72 is an endoplasmic reticulum luminal protein that is both a stress protein and a member of the protein disulfide isomerase family of proteins. Sequence analysis of ERp72 shows that it shares sequence identity with PDI (protein disulphide isomerase) at three discrete regions, as well as containing the ER retention signal KEEL at its C-terminus. ERp72 is shown to have three copies of sequence believed to be CGHC-containing active sites, which are also found in PDI. These CGHC sites are critical sites that are involved in the protein disulphide isomerization process. These results suggest that ERp72 is a newly described member of the family of CGHC-containing proteins that functions as a molecular chaperone.Specifica
| ERp72 | |
| Monoclonal | |
| 0.48 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P13667 | |
| PDIA4 | |
| ERp72 Fusion Protein Ag6873 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1D5F3 | |
| Unconjugated | |
| PDIA4 | |
| ER protein 70, ER protein 72, ERp 72, ERP70, ERp72, PDIA4, Protein disulfide isomerase A4 | |
| Mouse | |
| Protein G | |
| RUO | |
| 9601 | |
| -20°C | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto