Learn More
Estradiol benzoate, MedChemExpress
Product Code. 16442319
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16442319 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16442319 Supplier Medchem Express Supplier No. HYB1192
 Restricted Item
Spedizione stimata: 07-12-2026
MedChemExpress Estradiol Benzoate (β-Estradiol 3-benzoate), a prodrug of estradiol, acts as a steroid sex hormone. It exhibits mild anabolic and metabolic properties, and increases blood coagulability.
Estradiol Benzoate (β-Estradiol 3-benzoate), a prodrug of estradiol, acts as a steroid sex hormone. It exhibits mild anabolic and metabolic properties, and increases blood coagulability.
Specifications
Chemical Name or Material Estradiol benzoate
CAS 50-50-0
Color White
Purity Grade Notes Research
Molecular Formula C25H28O3
Quantity 1 g
Synonym β-Estradiol 3-benzoate 17β-Estradiol 3-benzoate
Solubility Information DMSO : 100 mg/mL (265.61 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES C[C@@]12[C@@H](O)CC[C@@]1([H])[C@]3([H])CCC4=C(C=CC(OC(C5=CC=CC=C5)=O)=C4)[C@@]3([H])CC2
Molecular Weight (g/mol) 376.49
Formula Weight 376.49
Percent Purity 98.04%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only