Learn More
TERUMO SR STRAIGHT FEP Intravenous Catheters SURFLO™
Click to view available options
Calibro dell'ago:
14 G
16 G
18 G
20 G
Lunghezza (sistema imperiale) ago:
1.25 in.
2 in.
2.5 in.
Dimensione della confezione:
pezzi 50
Descrizione
- The Surflo I.V. Catheters are made out of medical grade EFTE
- The materials of the Surflo™ I.V. Catheters are carefully selected to ensure a stable form during punctures under difficult conditions
- Short PIVC with an ETFE catheter tube and stainless steel internal needle
- Precisely tapered and siliconised catheter for smooth advancement of the catheter
- Catheter with X-ray contrast strips for higher patient safety when placement goes wrong
- The flashback chamber confirms needle tip entry into the vessel
- The large range in catheters and related obturators allows most appropriate choice of product for a wide range of applications
Specifica
Specifica
| Materiale | ETFE, Stainless Steel (Needle) |
| Colore | Green |
| Calibro dell'ago | 18 G |
| Senza lattice | Yes |
| Lunghezza (sistema imperiale) ago | 2 in. |
| Lunghezza (sistema metrico) | 51 mm |
| Product Type | IV Catheter |
| Monouso | Single-use |
| Tipo di collegamento | Leur Slip |
| Sterilità | Sterile |