Learn More
Ethoxycarbonyl isothiocyanate, 97%, Thermo Scientific Chemicals
72.90€ - 214.00€
Identificatori chimici
CAS | 16182-04-0 |
---|---|
Formula molecolare | C4H5NO2S |
Molecular Weight (g/mol) | 131.149 |
Numero MDL | MFCD00004814 |
InChI Key | BDTDECDAHYOJRO-UHFFFAOYSA-N |
Sinonimo | ethoxycarbonyl isothiocyanate, ethyl isothiocyanatoformate, carbethoxy isothiocyanate, ethoxycarbonylisothiocyanate, ethyl n-carbothioylcarbamate, ethyl n-sulfanylidenemethylidene carbamate, ethyl isothiocyanatidocarbonate, carbon isothiocyanatidic acid, ethyl ester, o-ethyl carbonisothiocyanatidate |
PubChem CID | 85320 |
IUPAC Name | ethyl N-(sulfanylidenemethylidene)carbamate |
SMILES | CCOC(=O)N=C=S |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
11384354
|
Thermo Scientific Alfa Aesar
L00911.06 |
5 g |
72.90€
5g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
11394354
|
Thermo Scientific Alfa Aesar
L00911.14 |
25 g |
214.00€
25g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
Ethoxycarbonyl isothiocyanate, is used as a building block in the synthesis of various heterocycles. It is also used for the synthesis of 1,2,4-oxadiazolidine-3,5-dione. It is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthoxycarbonyl isothiocyanate, is used as a building block in the synthesis of various heterocycles. It is also used for the synthesis of 1,2,4-oxadiazolidine-3,5-dione. It is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in chloroform and ether.
Notes
Moisture, Heat Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Identificatori chimici
16182-04-0 | |
131.149 | |
BDTDECDAHYOJRO-UHFFFAOYSA-N | |
85320 | |
CCOC(=O)N=C=S |
C4H5NO2S | |
MFCD00004814 | |
ethoxycarbonyl isothiocyanate, ethyl isothiocyanatoformate, carbethoxy isothiocyanate, ethoxycarbonylisothiocyanate, ethyl n-carbothioylcarbamate, ethyl n-sulfanylidenemethylidene carbamate, ethyl isothiocyanatidocarbonate, carbon isothiocyanatidic acid, ethyl ester, o-ethyl carbonisothiocyanatidate | |
ethyl N-(sulfanylidenemethylidene)carbamate |
Specifica
Ethoxycarbonyl isothiocyanate | |
1.11 | |
50°C (122°F) | |
C4H5NO2S | |
MFCD00004814 | |
606091 | |
ethoxycarbonyl isothiocyanate, ethyl isothiocyanatoformate, carbethoxy isothiocyanate, ethoxycarbonylisothiocyanate, ethyl n-carbothioylcarbamate, ethyl n-sulfanylidenemethylidene carbamate, ethyl isothiocyanatidocarbonate, carbon isothiocyanatidic acid, ethyl ester, o-ethyl carbonisothiocyanatidate | |
BDTDECDAHYOJRO-UHFFFAOYSA-N | |
ethyl N-(sulfanylidenemethylidene)carbamate | |
85320 | |
97% |
16182-04-0 | |
44°C to 46°C (10 mmHg) | |
Pungent | |
1.503 | |
UN2924 | |
Moisture sensitive | |
Soluble in chloroform and ether. | |
CCOC(=O)N=C=S | |
131.149 | |
131.15 |
Sicurezza e movimentazione
GHS H Statement
H301-H311-H314-H318-H226-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if inhaled.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P312-P330-P363-P370+P378q-P501c
H226-H301+H311+H331-H318-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II
missing translation for 'einecsNumber' : 240-318-9
missing translation for 'tsca' : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only