Learn More
Ethylmalonic acid, 97+%
Codice prodotto. p-7025304
Click to view available options
Quantità:
5 g
25 g
100 g
Dimensione della confezione:
100g
25g
5g
Codice prodotto. 11485566
Marca: Thermo Scientific Alfa Aesar B20715.06
Richiesta di formato massivo o personalizzato
CAS: 601-75-2 | C5H8O4 | 132.115 g/mol
Ethylmalonic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethylmalonic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in water, methanol (very faint turbidity), and ethanol.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 601-75-2
Formula molecolare C5H8O4
Molecular Weight (g/mol) 132.115
Numero MDL MFCD00002668
InChI Key UKFXDFUAPNAMPJ-UHFFFAOYSA-N
Sinonimo ethylmalonic acid, 2-ethylmalonic acid, propanedioic acid, ethyl, 1,1-propanedicarboxylic acid, alpha-carboxybutyric acid, malonic acid, ethyl, ethylmalonate, ethyl-malonic acid, unii-432nf49dfg, ethylpropanedioic acid
PubChem CID 11756
ChEBI CHEBI:741548
IUPAC Name 2-ethylpropanedioic acid
SMILES CCC(C(=O)O)C(=O)O
Specifica
Materiale o nome chimico Ethylmalonic acid
Punto di fusione 112°C to 114°C
Quantità 5 g
Beilstein 774334
Informazioni di solubilità Soluble in water,methanol (very faint turbidity),and ethanol.
Peso formulazione 132.12
Percent Purity 98%