Learn More
R&D Systems™ Etoposide
Click to view available options
Quantità:
100 μL
Dimensione della confezione:
100µL
Descrizione
- Use a manual defrost freezer and avoid repeated freeze-thaw cycles.
- Do not use past expiration date.
- Blocks the cell cycle in S-phase and G2-phase; induces apoptosis in normal and tumor cell lines; inhibits synthesis of the oncoprotein Mdm2 and induces apoptosis in tumor lines that overexpress Mdm2.
- Etoposide is provided in DMSO. The final reagent concentration is 10mM.
Specifica
Specifica
| Quantità | 100 μL |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |