Learn More
Evocalcet, MedChemExpress
Product Code. 16487418
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16487418 10 mg 10mg
16479897 5 mg 5mg
16453447 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16487418 Supplier Medchem Express Supplier No. HY17613
 Restricted Item
Spedizione stimata: 23-11-2026
MedChemExpress Evocalcet has an activating effect on calcium sensing receptor (CaSR) extracted from patent WO 2017061621 A1, compound A.
Evocalcet has an activating effect on calcium sensing receptor (CaSR) extracted from patent WO 2017061621 A1, compound A.
Specifications
Chemical Name or Material Evocalcet
CAS 870964-67-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H26N2O2
Quantity 10 mg
Synonym KHK7580
Solubility Information DMSO : 30 mg/mL (80.11 mM; Need ultrasonic and warming)
SMILES O=C(O)CC1=CC=C(N2C[C@@H](N[C@@H](C3=C4C=CC=CC4=CC=C3)C)CC2)C=C1
Molecular Weight (g/mol) 374.48
Formula Weight 374.48
Percent Purity 99.05%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less
Research purposes only