Learn More
EVSCO™ Three-Way, Two-Position Elliptic Valve
Click to view available options
Dimensioni dei tubi:
1/4 in.
3/8 in.
Unit Size:
Pezzo
Description
- Valves are easy to disassemble.
- Simply pull out the handle and push your stem through the bottom allowing you to clean and maintain valves in-line.
- Four-position valves are clearly marked with flow patterns to ensure correct handle position.
- Choose from FDA-grade PP or PVDF bodies.
- All valves have O-rings made of Viton, 316 SS handles, threaded connections and nonrestrictive orifice.
Specifications
Specifications
| Descrizione | Three-Way, Two-Position Elliptic Valve |
| Materiale | Polypropylene, Viton |
| Dimensioni dei tubi | 3/8 in. |
| Porta di uscita | 3/8 in. NPT(F) |