Learn More
Invitrogen™ EXOSC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA588182
Descrizione
Immunogen sequence: MAEPASVAAE SLAGSRARAA RTVLGQVVLP GEELLLPEQE DAEGPGGAVE RPLSLNARAC SRVRVVCGPG LRRCGDRLLV TKCGRLRHKE PGSGSGGGVY WVDSQQKRYV PVKGDHVIGI VTAKSGDIFK VDVGGSEPAS LSYLSFEGAT KRNRPNVQVG DLIYGQFVVA NKDMEPEMVC IDSCGRANGM GVIGQDGLLF KVTLGLIRKL LAPDCEIIQE VGKLYPLEIV FGMNGRIWVK AKTIQQTLIL ANILEACEHM TSDQRKQIFS RLAES; Positive Samples: A-431, HeLa, U-87MG, Raji, SGC-7901, Mouse brain, Rat brain; Cellular Location: Cytoplasm, Nucleus, nucleolus.
EXOSC3 is component of the exosome 3'->5' exoribonuclease complex and is required for the 3' processing of the 7S pre-RNA to the mature 5. 8S rRNA.
Specifica
| EXOSC3 | |
| Polyclonal | |
| Unconjugated | |
| Exosc3 | |
| 2310005D06Rik; AI593501; bA3J10.7; CGI-102; Exosc3; Exosome complex component RRP40; exosome complex exonuclease RRP40; exosome complex exonuclease RRP40; exosome complex component RRP40; exosome component 3; hRrp-40; hRrp40p; hypothetical protein LOC533245; LOW QUALITY PROTEIN: exosome complex component RRP40; p10; PCH1B; Ribosomal RNA-processing protein 40; RP11-3J10.8; RRP40; Rrp40p | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 313243, 51010, 66362 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.42 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q7TQK4, Q9NQT5 | |
| Exosc3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-275 of human EXOSC3 (NP_0571262). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.