missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EXOSC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA530610
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Recommended positive controls: A431, Jurkat, Raji. Predicted reactivity: Mouse (86%), Rat (83%), Rhesus Monkey (97%), Chimpanzee (98%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
EXOSC3 is component of the exosome 3'->5' exoribonuclease complex and is required for the 3' processing of the 7S pre-RNA to the mature 5. 8S rRNA.
Specifica
| EXOSC3 | |
| Polyclonal | |
| Unconjugated | |
| Exosc3 | |
| 2310005D06Rik; AI593501; bA3J10.7; CGI-102; Exosc3; Exosome complex component RRP40; exosome complex exonuclease RRP40; exosome complex exonuclease RRP40; exosome complex component RRP40; exosome component 3; hRrp-40; hRrp40p; hypothetical protein LOC533245; LOW QUALITY PROTEIN: exosome complex component RRP40; p10; PCH1B; Ribosomal RNA-processing protein 40; RP11-3J10.8; RRP40; Rrp40p | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51010 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9NQT5 | |
| Exosc3 | |
| Recombinant protein encompassing a sequence within the center region of human EXOSC3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto