missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXOSC3 Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Marca: Abnova PAB30057.100uL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Rabbit polyclonal antibody raised against synthetic peptide of human EXOSC3.
- High specificity
- High reactivity
Specifica
| EXOSC3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of human EXOSC3. | |
| In PBS (2% sucrose, 0.09% sodium azide). | |
| CGI-102/MGC15120/MGC723/RP11-3J10.8/RRP40/Rrp40p/bA3J10.7/hRrp40p/p10 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 51010 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-8 ug/mL) Western Blot (0.2-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| NM_016042 | |
| EXOSC3 | |
| A synthetic peptide corresponding to internal region of human EXOSC3. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only (RUO)
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur