Learn More
EXPH5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 22199-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The function of this protein remains unknown.Specifica
| EXPH5 | |
| Polyclonal | |
| Unconjugated | |
| EXPH5 | |
| exophilin 5, EXPH5, KIAA0624, SLAC2 B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23086 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8NEV8 | |
| EXPH5 | |
| EXPH5 Fusion Protein Ag17449 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |