Learn More
Ismatec™ Extension Tubing, Tygon 2001
Click to view available options
Dimensioni dei tubi:
I.D. 2.79 mm
Dimensione della confezione:
Pezzo
Descrizione
- Transparent.
- Superior pump life.
- Especially designed for use MEK and other aggressive solvents.
- Plasticizer and oil-free.
- Smooth inner-bore.
- Low sorption maintains fluid and tube integrity.
- Does not impart anything into the pumping medium.
- No release of hazardous materials when properly incinerated
Specifica
Specifica
| Certificazione/Conformità | FDA 21 CFR 177.2600;FDA Approved for contact with foods |
| Autoclavabile | Autoclavable |
| Diametro (sistema metrico) interno | 2.79 mm |
| Materiale | Tygon MHSL 2001 |
| Overall Length (Metric) | 10 m |
| Min. Operating Temperature (Metric) | −73°C |
| Quantità confezione | 1 Roll |
| Resistance Properties | Acids: Excellent, Alkaline solutions: Excellent, Solvents: Good, Pressure: Good, Vacuum: Good, Viscous media: Excellent, Sterile media: Good |
| Dimensioni dei tubi | I.D. 2.79 mm |
| Metodo di sterilizzazione | Autoclaveable with steam, 30 minutes at 1 bar (15 psi) and 141°C (250°F). Gas sterilization with ethylene oxide. Sterilization with radiation up to 2.5 mrad. |
| Vedi altri risultati |