Learn More
Ismatec™ Extension Tubing, Tygon ST
Codice prodotto. 10027331
Click to view available options
Dimensioni dei tubi:
I.D. 0.19 mm
I.D. 0.38 mm
I.D. 0.57 mm
I.D. 0.89 mm
I.D. 1.14 mm
I.D. 1.65 mm
I.D. 1.85 mm
Dimensione della confezione:
10m
Pezzo
Codice prodotto. 10027331
Marca: Ismatec™ 0550240
For use with 2- and 3-stop color-coded tubing.
  • Inexpensive, all-around tubing for general laboratory applications.
  • Transparent
  • Resistant to almost all inorganic chemicals
  • Tasteless
  • Smooth, polished inner wall
  • Low gas permeability
  • Non-aging and non-oxidizing
  • High dialectric constant
Specifica
Certificazione/Conformità FDA 21 CFR175.300
Autoclavabile Autoclavable
Diametro (sistema metrico) interno 1.85 mm
Materiale Tygon ST R-3603, R-3607
Overall Length (Metric) 10 m
Da utilizzare con (applicazione) General laboratory applications, 2- and 3-stop color-coded tubing.
Min. Operating Temperature (Metric) −50°C
Quantità confezione 1 Reel
Resistance Properties Acids: Good, Alkaline solutions: Good, Solvents: Not rec., Pressure: Fair, Vacuum: Good, Viscous media: Excellent, Sterile media: Limited
Dimensioni dei tubi I.D. 1.85 mm
Metodo di sterilizzazione Autoclavable with steam, 30 minutes at 1 bar (15 psi) and 121°C (250°F); tubing will appear milky.Gas sterilization with Ethylene oxide.Not recommended for sterilization with radiation.
Colore Trasparente
Max. Operating Temperature (Metric) 74°C