Learn More
Lenz Laborglasintrumente Extractors Acc. to Soxhlet with Dimroth Condenser
Click to view available options
Capacità (sistema metrico):
70 mL
100 mL
150 mL
300 mL
500 mL
Dimensione della confezione:
Pezzo
Descrizione
- The method allows an effective separation with relatively small amounts of extracting solvents.
- Extractor in compact and robust design, with flask connection NS 29/32
- Made of DURAN™ tubing
- Dimroth condenser with thread GL 14
- Round bottom flask
Specifica
Specifica
| Capacità (sistema metrico) | 150 mL |
| Materiale | Duran Glass |
| Giunto rastremato standard | NS 45/40 |