Learn More
Lenz Laborglasintrumente Extractors Acc. to Soxhlet with Dimroth Condenser
Codice prodotto. p-7222538
Click to view available options
Capacità (sistema metrico):
70 mL
100 mL
150 mL
300 mL
500 mL
Dimensione della confezione:
Pezzo
Codice prodotto. 10382761
Marca: Lenz Laborglasintrumente 05360025
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • The method allows an effective separation with relatively small amounts of extracting solvents.
  • Extractor in compact and robust design, with flask connection NS 29/32
  • Made of DURAN™ tubing
  • Dimroth condenser with thread GL 14
  • Round bottom flask
Specifica
Capacità (sistema metrico) 100 mL
Materiale Duran Glass
Giunto rastremato standard NS 29/32