missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ EZ-Link™ Hydrazide-PEG4-Desthiobiotin, No-Weigh™ Format
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Quantità:
5 x 1 mg
Dimensione della confezione:
pezzi 5
Descrizione
- Pegylated, carbonyl-reactive labeling reagent whose biotin-like group is elutable from streptavidin, enabling its use for cell surface glycoprotein labeling and purification.
- Desthiobiotin - analog of biotin allows easy elution from streptavidin, an ideal feature for affinity purification applications
- Glycoprotein labeling - tag glycosylated proteins at sialic acid residues to enable high recovery in pull-down assays with streptavidin beads
- Cell surface labeling - tag and isolate cell surface glycoproteins; reagent does not permeate membranes of whole cells
- Aldehyde-reactive - reacts with aldehydes formed by periodate-oxidation of sugar groups
- Hydrazide-activated - perform reactions at pH 4 to 6 in buffers such as sodium acetate
- Pegylated - spacer arm contains a hydrophilic, 4-unit, polyethylene glycol (PEG) group
- Enhances solubility - pegylation imparts water solubility to the biotinylated molecule, helping to prevent aggregation of biotinylated antibodies stored in solution
Specifica
Specifica
| Product Type | Hydrazide-PEG4-Desthiobiotin |
| Quantità | 5 x 1 mg |
| Content And Storage | Store at 4°C. |
| Labeling Method | Chemical Labeling |
| Shipping Condition | Wet Ice |
| Linea di prodotti | EZ-Link™ |
| Permeabilità celle | Cell-Impermeant |
| Chemical Reactivity | Carboxylic Acid, Ketone, Aldehyde |
| Reactive Moiety | Amine, Hydrazide |
| Solubility | DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water |
| Vedi altri risultati |
For Research Use Only. Not for use in diagnostic procedures.
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto