Learn More
Ezrin Antibody (CL2360) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Novus Biologicals NBP2-36761PE
Codice prodotto. 30297759
  • 885.00€ / 0.10mL
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
Ezrin Monoclonal antibody specifically detects Ezrin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifica
Specifica
Ezrin
Monoclonal
PE
CVIL, CVL, Cytovillin, cytovillin 2, DKFZp762H157, EZR, ezrin, FLJ26216, p81, VIL2MGC1584, villin 2 (ezrin), villin-2
This antibody was developed using a recombinant protein derived from P15311, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cell Cycle and Replication, Phospho Specific, Signal Transduction
7430
Store at 4⪚C in the dark.
IgG2b
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL2360
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified