missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM3C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14247-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
FAM3C is a 227 amino acid secretory protein that belongs to a novel cytokine-like family with sequence similarity to FAM3 family. FAM3C is a signaling cytokine with a GG domain, hydrophobic leader sequence and an N-terminal signal peptide. FAM3C has a cytokine activity and shows a change in expression in pancreatic cancer-derived cells. It is ubiquitously expressed in most of the tissues. Reports suggest that FAM3C plays an important role in inner ear development and may thus be involved in cell differentiation and proliferation during inner ear embryogenesis.Specifica
| FAM3C | |
| Polyclonal | |
| Unconjugated | |
| FAM3C | |
| FAM3C, GS3786, ILEI, Interleukin like EMT inducer, Protein FAM3C | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10447, 27999, 312159 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q810F4, Q91VU0, Q92520 | |
| FAM3C | |
| FAM3C Fusion Protein Ag5516 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto