missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fascin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14384-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Human fascin is a highly conserved actin-bundling protein. Fascin, encoded by the human homolog for sn (hsn) gene, has been localized to microspikes and stress fibers of cultured cells where it is thought to be involved in the formation of microfilament bundles. It is expressed predomitly in dendritic cells. Lymphoid cells, myeloid cells and plasma cells are negative. However. Reed Sternberg cells in Hodgkin′s lymphoma are positive for fascin staining. Epstein-Barr virus may induce expression of fascin in B cells.Specifica
| Fascin | |
| Polyclonal | |
| Unconjugated | |
| FSCN1 | |
| 55 kDa actin bundling protein, FAN1, Fascin, Fascin 1,Fascin, FSCN1, HSN, p55, Singed like protein, SNL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14086, 6624 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q16658, Q61553 | |
| FSCN1 | |
| Fascin Fusion Protein Ag5816 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto