Learn More
Thermo Scientific™ FastDigest™ NheI: PROMO
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Quantità:
1 each
Dimensione della confezione:
50 reazioni
Descrizione
- 100% activity of all FastDigest enzymes in the universal buffer
- 100% buffer compatibility with downstream applications
- 100% buffer compatibility with downstream applications
- Direct loading on gels
- Direct loading on gels
- 176 FastDigest enzymes available
Specifica
Specifica
| Optimal Reaction Temperature | 37°C |
| Quantità | 1 each |
| Sito di taglio | G^CTAGC |
| Product Type | Restriction Enzyme |
| Product Line | FastDigest |