Learn More
Thermo Scientific™ FastRuler DNA Ladders, ready-to-use
GREENER_CHOICE
Codice prodotto. p-200113685 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Intervallo dimensioni:
10 to 200 bp
100 to 5000 bp
50 to 1500 bp
500 to 10000 bp
Dimensione della confezione:
1000µL
pezzi 2
Codice prodotto. 10410280
Marca: Thermo Scientific™ SM1233
Thermo Scientific FastRuler DNA Ladders are engineered for fast and convenient sizing and approximate quantification of double stranded DNA fragments.
Thermo Scientific FastRuler DNA Ladders is a fast and convenient tool for double-stranded DNA sizing during electrophoresis. It's specifically engineered for rapid sizing and approximate quantification of DNA on both conventional agarose gels and high-throughput E-Gels (in 48- and 96-well formats).
The ladder is a mixture of five blunt-ended chromatography-purified individual DNA fragments. This allows you to achieve precise results faster, saving valuable lab time.
Highlights
  • Variety of available ranges (from ultra-low to high- range)
  • Fast separation (8 to 14 min)
  • Short separation distance (10 to 20 mm)
  • Sharp bands, easy-to-remember fragment sizes and quantities
  • Ready-to-use: premixed with 6X MassRuler DNA Loading Dye for direct loading
  • Stable at room temperature for up to 24 months
Sustainability features
Ready-to-use FastRuler DNA ladders can be stored at room temperature for up to 24 months. The ladder is shipped at ambient temperature, using optimized right-sized packaging and 100% recyclable packaging reducing waste.
TRUSTED_SUSTAINABILITY
Specifica
Gel Type Agarose
Label or Dye Xylene Cyanol FF, Orange G
Product Type DNA Ladder
Ready to Load Yes
Intervallo dimensioni 10 to 200 bp
Compatibilità gel Agarose Gels
Green Features Sustainable packaging
Linea di prodotti FastRuler
Quantità 2 x 500 μL
For Research Use Only. Not for use in diagnostic procedures.