missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO36 Monoclonal Antibody (3D3), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA524517
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Peptide Sequence: HLQGQTALIF GARILDYVIN FCKGKFDFLE RLSDDLLLTI ISYLDLEDIA RLCQTSHRFA KLCMSDKLWE QIVQSTCDTI TPDVRALAED TGWRQLFFTN
Members of the F-box protein family, such as FBXO36, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).Specifica
FBXO36 | |
Monoclonal | |
Unconjugated | |
FBXO36 | |
0610008D19Rik; 1110020F21Rik; 2410002G19Rik; D1Ertd757e; F-box only protein 36; F-box protein 36; Fbx36; Fbxo36 | |
Mouse | |
Protein A | |
RUO | |
130888 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
3D3 | |
PBS with no preservative; pH 7.4 | |
Q8NEA4 | |
FBXO36 | |
FBXO36 (NP_777559, 66 a.a. ∽ 165 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |