Learn More
FDC Antibody (CNA.42), Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP1-43118-0.025MG
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
FDC Monoclonal specifically detects FDC in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifica
| FDC | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| cardiomyopathy, dilated 1B (autosomal dominant), CMPD1, FDC | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 1218 | |
| Store at 4C. Do not freeze. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| CNA.42 | |
| Unconjugated | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| CMD1B | |
| The immunogen for this antibody was follicular dendritic cells. | |
| 0.025 mg | |
| Primary | |
| Human | |
| IgM |
Titolo del prodotto
For Research Use Only