Learn More
Value Plastics Female Luer Thread Style to UNF Threaded Adapter Fittings, Straight
Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Dimensioni filo:
1/4-28 UNF(M)
10-32 UNF(M)
Ingresso:
Female Luer with 1/4 in. Hex
Female Luer with 5/16 in. Hex
Materiale (guarnizione):
Nylon
Polycarbonate
Polypropylene
Dimensione della confezione:
pezzi 25
Descrizione
Quick and easy assembly and disassembly without the need for tools. Thread design provides a secure closure—preventing contamination. Conforms to ISO-594-1 and ISO 594-2 standards
Specifica
Specifica
| Certificazione/Conformità | Non-DEHP;Non-Phthalate;RoHS |
| Quantità confezione | 25/Pk. |
| Ingresso | Female Luer with 5/16 in. Hex |
| Dimensioni filo | 1/4-28 UNF(M) |
| Materiale (guarnizione) | Nylon |