Learn More
Fenofibrate, TRC
Product Code. 17943515
Change view
Click to view available options
Quantity:
100 mg
1 g
25 g
Unit Size:
100mg
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17943515 100 mg 100mg
30134127 1 g 1g
30135200 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17943515 Supplier Toronto Research Chemicals Supplier No. TRCF248640100MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 49562-28-9
Molecular Formula C20 H21 Cl O4
Molecular Weight (g/mol) 360.83
Synonym Fenofibrate, 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoic acid 1-methylethyl ester, Ankebin, Antara, Clorofibrate, Elasterin, Fenobrate, Fenogal, Fenotard, Isopropyl 2-[p-(p-chlorobenzoyl)phenoxy]-2-methylpropionate, LF 178, Lipanthyl, Lipantil, Lipidil, Lipidil Supra, Lipirex, Lipoclar, Lipofene, Liposit, Lipsin, NSC 281319, Nolipax, Procetofen, Procetofene, Procetoken, Protolipan, Secalip, TriCor
IUPAC Name propan-2-yl 2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoate
SMILES CC(C)OC(=O)C(C)(C)Oc1ccc(cc1)C(=O)c2ccc(Cl)cc2
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C20H21ClO4/c1-13(2)24-19(23)20(3,4)25-17-11-7-15(8-12-17)18(22)14-5-9-16(21)10-6-14/h5-13H,1-4H3
Formula Weight 360.1128
Percent Purity >95%
Chemical Name or Material Fenofibrate
Grade HPLC