Learn More
Hirschmann™ Discharge Tube Unit with Luer-Lock Connection
Codice prodotto. 10293621
Click to view available options
:
For use with Ceramus, Opus dos, Battery-Drive ejection unit dispenser
Codice prodotto. 10293621
Marca: Hirschmann™ 9313029
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Dispensing nozzle with FEP-tubing and Luer-Lock-Connection for use with ceramus™ and opus™ dispenser.
  • Guided tubing
  • Screw-coupled
  • With Luer-Lock connector for filtration
Specifica
Product Type Discharge Tube Unit
Materiale FEP, PFA
Da utilizzare con (apparecchiatura) Ceramus, Opus dos, Battery-Drive ejection unit dispenser
Include luer-lock connector