Learn More
Cole-Parmer FEP Tubing
Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Diametro (sistema imperiale) interno:
1/16 in.
1/2 in.
1/8 in.
3/16 in.
3/32 in.
5/32 in.
7/16 in.
Diametro (sistema imperiale) esterno:
1/2 in.
1/4 in.
1/8 in.
3/16 in.
5/32 in.
9/16 in.
missing translation for 'unitSize'
15m
Description
Made of 100% virgin-grade fluorinated ethylene propylene (FEP). Excellent chemical and thermal resistance. Great optical clarity—easily view your sample in use. Sterilize by autoclave, EtO, and chemical sterilants. Complies with FDA 21 CFR 177.1550, USP Class VI, NSF 51, NSF 61, NSF 372, and ASTM D2116. Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection
Spécification
Spécification
| Certificazione/Conformità | ADCF;FDA;NSF-51;NSF-61;RoHS;USP |
| Diametro (sistema imperiale) interno | 1/8 in. |
| Diametro (sistema imperiale) esterno | 3/16 in. |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu