Learn More
Thermo Scientific™ Acc65I (Asp718I) (10 U/μL)
Codice prodotto. p-4521435 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Quantità:
1,000 units
5,000 units
Dimensione della confezione:
1000 unità
5000 unità
Codice prodotto. 10232680
Marca: Thermo Scientific™ ER0901
Edge
Cut at G^GTACC sites with Acc65I (Asp718I) restriction enzyme, which performs best at 37°C in O buffer (Isoschizomers: Asp718I).
Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
Highlights
  • Superior quality – stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Applications
  • Molecular cloning
  • Restriction site mapping
  • Genotyping
  • Southern Blot
  • Restriction fragment length polymorphism (RFLP)
  • SNP
TRUSTED_SUSTAINABILITY
Specifica
Concentrazione 10 U/μL
Methylation Sensitivity CpG Methylation-Sensitive, Not CpG Methylation-Sensitive, Not Dam Methylation-Sensitive
Enzyme Acc65I (Asp718I)
Compatible Buffer 10X Buffer O
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE No
Quantità 1,000 units
Product Type Restriction Enzyme
Research Category Traditional Cloning