Learn More
Thermo Scientific™ High Fidelity PCR Enzyme Mix Not available in US
Codice prodotto. 10212650 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Quantità:
100U
500U
Dimensione della confezione:
100 unità
500 unità
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
10212650 100U 100 unità
10191370 500U 500 unità
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 10212650 Fornitore Thermo Scientific™ N. del fornitore K0191
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
A Blend of Taq DNA Polymerase and a thermostable DNA polymerase with proofreading activity.
Thermo Scientific High Fidelity PCR Enzyme Mix is a unique blend of Taq DNA Polymerase and a thermostable DNA polymerase with proofreading activity. The two enzymes synergistically generate longer PCR products with greater yield and fidelity than Taq DNA Polymerase alone. Thus, fidelity of the High Fidelity PCR Enzyme Mix exceeds more than four times the fidelity of Taq DNA Polymerase. In addition, the enzyme mix can generate long amplicons up to 20 kb with viral DNA templates and up to 10 kb with genomic DNA templates. Such long amplicons are often difficult to generate with a single DNA polymerase.
The two enzymes synergistically generate longer PCR products with greater yield and fidelity than Taq DNA Polymerase alone. Thus, fidelity of the High Fidelity PCR Enzyme Mix exceeds more than four times the fidelity of Taq DNA Polymerase. In addition, the enzyme mix can generate long amplicons up to 20kb with viral DNA templates and up to 10kb with genomic DNA templates. Such long amplicons are often difficult to generate with a single DNA polymerase.
  • Unique blend of two thermostable DNA polymerases
  • Four-times greater accuracy than Taq DNA polymerase
  • Long amplification products:—up to 20kb with viral DNA as template, —up to 10kb with genomic DNA as template
  • High yields
  • Robust
Recommended for:
Generation of PCR products for purposes of cloning, expression, site-directed mutagenesis; High yield PCR; Long range PCR; RT-PCR.
Note:
High Fidelity PCR Enzyme Mix generates both blunt-ended and 3'-dA tailed PCR products. Blunting of PCR product prior to cloning is highly recommended. For efficient PCR product cloning, use the CloneJET PCR Cloning Kit.
TRUSTED_SUSTAINABILITY
Specifica
Quantità 100U
In certain countries use of this product is covered by patents. Purchase of product in these countries includes non-transferable, limited license for using only this amount of product for the purchaser's own internal research.
This product is licensed under one U.S. Patent No. 5,436,149 or corresponding foreign patents owned by Takara Bio, Inc.