Learn More
Coverguard™ FFP2 NR D Face Mask
Descrizione
- PPE Cat. III FFP2 NR D
- Disposable
- Electrostatic filter media
- Sliding elastic band for better adjustment
- Latex-free, non-woven polypropylene
- Adjustable nose clip
Specifica
Specifica
| Certifications/Compliance (Europe) | CE PPE Category III FFP2 NR D, EN 149:2001 + A1:2009 |
| Descrizione | FFP2 Disposable Shell Mask, Welded Outer Contour |
| Monouso | Single-use |
| With Exhalation Valve | No |
| Da utilizzare con (apparecchiatura) | Chemical and Pharmaceutical Industries, Light Industries, Heavy and Process Industries, Construction Finishings, Building Industry, Infrastructure |
| Tipo di protezione nasale | Adjustable Nose Clip with PU Foam Nose Pad |
| Resistente a | Non-Toxic Solid and Liquid Aerosols |
| Sterilità | Non-sterile |
| Stile | Shell Mask |