Learn More
Coverguard™ FFP2 NR D SL VO-GA Half Mask for Organic Vapors and Acids, Blue with Exhalation Valve
Click to view available options
Descrizione:
Blue FFP2 Half Mask with Shell and Carbon Valve
Sterilità:
Non-sterile
Resistente a:
Aerosols and Low Toxic Dust
Dimensione della confezione:
pezzi 120
Descrizione
- PPE Cat. III FFP2 NR D Organic vapors, Gas and Acids
- Filters bad odors
- Exhalation valve with active carbon reduces fog build-up inside the mask
- Non-woven polypropylene
- Adjustable aluminum nose clip with PU foam total nose seal
- Specific design gives limited breathing resistance thanks to the large surface of the filtering material
Specifica
Specifica
| Certifications/Compliance (Europe) | CE PPE Cat. III, FFP2, EN 149:2001 + A1:2009 |
| Descrizione | Blue FFP2 Half Mask with Shell and Carbon Valve |
| Monouso | Single-use |
| With Exhalation Valve | Sì |
| Da utilizzare con (apparecchiatura) | Heavy & Process Industries, Chemical & Pharmaceutical Industries, Light Industries, Infrastructure, Building Industry, Construction - Finishing Work, DIY |
| Tipo di protezione nasale | Adjustable Aluminum Nose Clip, PU Foam Nose Pad |
| Resistente a | Aerosols and Low Toxic Dust |
| Forma | Half Mask |
| Sterilità | Non-sterile |