Learn More
Invitrogen™ FGF14 Recombinant Rabbit Monoclonal Antibody (107)

Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen™ MA530426
Descrizione
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human FGF14/SCA27.
FGF14 is involved in nervous system development and function.
Specifica
| FGF14 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q92915 | |
| FGF14 | |
| Recombinant Human FGF14/SCA27 protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 107 | |
| Unconjugated | |
| FGF14 | |
| bA397O8.2; FGF; FGF homologus factor 4b; Fgf14; FGF-14; FHF4; FHF-4; Fibroblast growth factor; Fibroblast growth factor 14; fibroblast growth factor 14, isoform b; fibroblast growth factor homologous factor 4; m 1B; mFHF-4(1B); SCA27 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 2259 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.