Learn More
FGL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11827-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Fibrinogen-like protein 2 (Fgl2), also known as fibroleukin, is secreted by T cells and is involved in diseases in which thrombosis plays a pivotal role,such as virus-induced fulmit hepatitis, fetal loss syndrome and xenograft rejection. Constitutively xpressed in cytotoxic T cells, Fgl2 exerts immunosuppressive effects on both T cell proliferation and dendritic cell maturation.Fgl2 is a serine protease and directly cleaves prothrombin to thrombin. Fgl2 functions in the pathogenesis of diseases including viral-induced hepatitis and Th1 cytokine-induced fetal loss syndrome.Specifica
| FGL2 | |
| Polyclonal | |
| Unconjugated | |
| FGL2 | |
| FGL2, FGL-2, fibrinogen like 2, Fibrinogen like protein 2, Fibroleukin, pT49, T49 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10875, 14190, 84586 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P12804, Q14314 | |
| Fgl2 | |
| FGL2 Fusion Protein Ag2395 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |