Learn More
Thermo Scientific Chemicals Fibroblast Growth Factor 10, human
Descrizione
Plays an important role in the regulation of embryonic development, cell proliferation and cell differentiation. Required for normal branching morphogenesis. May play a role in wound healing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Peso molecolare | 19.3 kd |
| Quantità | 25μg |
| Imballaggio | Plastic bottle |
| Forma | Lyophilized |